Learn More
Medchemexpress LLC Phenacetin-d5 (acetophenetidin-d5) | 69323-74-6 | 99.5% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0476S5MG
Encompass_Preferred
Phenacetin-d5 is the deuterium-labeled version of Phenacetin. Phenacetin (Acetophenetidin) is an analgesic/antipyretic agent that is non-opioid. It acts as a selective COX-3 inhibitor and is used as a probe for cytochrome P450 enzymes CYP1A2 in human liver microsomes and in rats. Deuteration, the incorporation of stable heavy isotopes of hydrogen into drug molecules, has shown potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06718