Learn More
Medchemexpress LLC Phenacetin (Standard) | 62-44-2 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0476R
Encompass_Preferred
Phenacetin (Standard) is an analytical standard of Phenacetin, intended for research and analytical applications. It is a non-opioid analgesic/antipyretic agent that acts as a selective COX-3 inhibitor and is used as a probe of cytochrome P450 enzymes CYP1A2. This product is for research use only.
  • Analytical standard for research and analytical applications
  • Non-opioid analgesic/antipyretic agent
  • Selective COX-3 inhibitor
  • Used as a probe of cytochrome P450 enzymes CYP1A2
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-50621