Learn More
Medchemexpress LLC Phenanthrene | 85-01-8 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB172710MM/1ML
Encompass_Preferred
Phenanthrene is an orally active polycyclic aromatic hydrocarbon (PAH) commonly used to detect or assess PAH pollution in the environment. It induces inflammation, oxidative stress, and apoptosis.
  • Reduces mitochondrial membrane potential.
  • Induces oxidative stress and inflammation.
  • Can cause liver damage and promote hepatocyte apoptosis.
  • Supplied as a 10 mM solution in DMSO.
Catalog No. 50-004-13315