Learn More
Medchemexpress LLC Phenanthrene (Standard) | 85-01-8 | 99.6% | 1 G

Supplier: Medchemexpress LLC HYB17271G
Phenanthrene (Standard) is an analytical standard used for research and analytical applications. This orally active polycyclic aromatic hydrocarbon (PAH) induces inflammation, oxidative stress, and apoptosis, and is commonly used to detect or assess PAH pollution in the environment.
- Analytical standard grade, supplied for assay
- Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
- Molecular weight: 178.23
- Formula: C14H10
- CAS no.: 85-01-8
- Appearance: Solid
- Color: White to off-white
- Ships at room temperature in continental US; may vary elsewhere
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.