Learn More
Medchemexpress LLC Phenanthrene (Standard) | 85-01-8 | 99.6% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1727R100MG
Encompass_Preferred
Phenanthrene (Standard) is an analytical standard and reference compound for research and analytical applications. This polycyclic aromatic hydrocarbon (PAH) induces inflammation, oxidative stress, and apoptosis. It is also commonly used to detect or assess PAH pollution in the environment.
  • Analytical standard for research and analytical applications
  • Reference standard for assay
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Commonly used in HPLC, GC, and MS
  • Induces inflammation, oxidative stress, and apoptosis
  • Used to detect or assess PAH pollution in the environment
Catalog No. 50-004-11923