Learn More
Medchemexpress LLC Phenanthrene (Standard) | 85-01-8 | 99.6% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB1727R50MG
Encompass_Preferred
Phenanthrene (Standard) is an analytical standard of Phenanthrene, intended for research and analytical applications. This orally active polycyclic aromatic hydrocarbon (PAH) can induce inflammation, oxidative stress, and apoptosis, and is commonly used to detect or assess PAH pollution in the environment.
  • Analytical standard grade
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Utilized in studies related to inflammation, oxidative stress, and apoptosis
  • Applied in assessing PAH pollution
Catalog No. 50-004-12534