Learn More
Medchemexpress LLC Phenanthriplatin | 1416900-51-0 | 98.06% | 503.80 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY139813
Encompass_Preferred
Phenanthriplatin is a monovalent platinum(II)-based complex (CAS: 1416900-51-0, MW: 503.80) with significant cytotoxicity against cancer cells, making it a potent anti-cancer agent. It is supplied as a solid, white to off-white appearance.
  • Monovalent platinum(II)-based complex
  • Exhibits significant cytotoxicity against cancer cells
  • Potent anti-cancer agent
Catalog No. 50-003-51606