Learn More
Medchemexpress LLC Phenanthriplatin | 1416900-51-0 | 98.1% | 503.80 | 50 MG

Supplier: Medchemexpress LLC HY13981350MG
Phenanthriplatin is a monovalent platinum(II)-based complex that exhibits significant cytotoxicity against cancer cells. It is for research use only and not sold to patients.
- Monovalent platinum(II)-based complex
- Exhibits significant cytotoxicity against cancer cells
- Molecular weight: 503.80
- Formula: C13H13ClN4O3Pt
- Appearance: Solid
- Color: White to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.