Learn More
Medchemexpress LLC Phenanthro[1,2-b]furan-11(2H)-one, 1,10-dihydro-10-hydroxy-1,6-dimethyl-10-(2-oxopropyl)- | 189308-08-5 | 99.7% | 1 MG

Supplier: Medchemexpress LLC HY1229171MG
Danshenol A is an abietane-type diterpenoid research compound and a potent aldose reductase inhibitor (IC50 = 0.1 μM) with reported antioxidant, anti-inflammatory, and antitumor activities. It is used in cardiovascular and inflammation-related research where ROS protection is studied.
- High purity (99.7%).
- Soluble in DMSO at ≥ 12.5 mg/mL; in vivo formulations reported at ≥ 1.25 mg/mL.
- Storage: 4°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
- Suitable for biochemical assays and small-scale in vivo studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.