Learn More
Medchemexpress LLC Phenazine-1-carboxylic acid | 2538-68-3 | 100.0% | 224.21 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY3303750MG
Encompass_Preferred
Phenazine-1-carboxylic acid is an antifungal agent that also exhibits anticancer activity by inducing apoptosis in cancer cells through the regulation of ROS generation. It can upregulate the expression of IL-8 and ICAM-1 while inhibiting the release of RANTES and MCP-1, indicating potential immunomodulatory effects. It is valuable for research in anti-infection, anticancer, and immune response modulation.
  • Antifungal agent
  • Exhibits anticancer activity
  • Induces apoptosis in cancer cells through ROS generation
  • Upregulates IL-8 and ICAM-1 expression
  • Inhibits release of RANTES and MCP-1
  • Antineuroinflammatory activity
  • Cytotoxicity against HCT-116 cells
  • Inhibits proliferation in DU145 cells
  • Significantly prevented V. anguillarum infection in zebrafish embryos
Catalog No. 50-004-04284