Learn More
Medchemexpress LLC Phenazine | 92-82-0 | 50g
SDP
Supplier:  Medchemexpress LLC HYW01647750G
Encompass_Preferred
Phenazine is an electron shuttles which modulates the redox state of cells and the downstream gene expression related to biofilm formation and bacterial survival Phenazines is a biocontrol agents which affects the growth of plants and induces the systemic resistance in plants Phenazine maintains NAD /NADH balance[1][2]
Catalog No. 50-002-99360