Learn More
Crescent Chemical Co Inc PHENAZINE-METHOSULFATE PURE
SDP
Supplier:  Crescent Chemical Co Inc SE32030.01
Encompass_Preferred
Phenazine-methosulfate pure Electron coupler, transfers electrons from NADH to tetrazolium salts, e.g. MTT, and thus makes NAD reductions visible. Size - 1G Storage Conditions - +2 °C TO +8 °C Catalog Number - 32030.01 CAS 299-11-6
Catalog No. 50-247-999