Learn More
Apexbio Technology LLC Phenazopyridine HCl 136-40-3 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B18125.1
Encompass_Preferred
Phenazopyridine HCl (CAS 136-40-3) is a small-molecule antagonist with competitive inhibitory activity (IC50 145 M) It exerts local analgesic effects primarily through topical action on the urinary tract mucosa thus alleviating pain symptoms associated with bladder and urethral irritation In experimental research phenazopyridine HCl is utilized in models investigating pain transmission pathways urinary tract epithelial responses and inflammatory modulation Based on these pharmacological properties phenazopyridine HCl holds research potential in the study of urinary tract inflammation epithelial permeability nerve differentiation and in assays evaluating analgesic activity in urological and neurobiological research
Catalog No. 50-000-96496