Learn More
Medchemexpress LLC Phenelzine (sulfate) | 156-51-4 | 99.96% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB1018A10MM/1ML
Encompass_Preferred
Phenelzine sulfate is an antidepressant agent that acts as an irreversible and orally active monoamine oxidase (MAO-A and MAO-B) inhibitor. It also inhibits GABA transaminase, primary amine oxidase (PrAO), and sequesters reactive aldehydes. This compound suppresses oxidative stress and lipogenesis, and elevates neurotransmitters like serotonin, norepinephrine, and dopamine. It is intended for research use only.
  • Antidepressant agent and MAO-A/MAO-B inhibitor
  • Inhibits GABA transaminase and primary amine oxidase (PrAO)
  • Sequestering reactive aldehydes
  • Inhibits LSD1 (Ki: 5.6 μM)
  • Suppresses oxidative stress and lipogenesis
  • Elevates serotonin, norepinephrine, and dopamine levels
  • Potential in neurological, metabolic, and cancer diseases
Catalog No. 50-004-07488