Learn More
Medchemexpress LLC Pheneturide | 90-49-3 | 99.84% | 206.24 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY111177R10MG
Encompass_Preferred
Pheneturide (Standard) is the analytical standard of Pheneturide, intended for research and analytical applications. It is a decarboxylation product of Phenobarbital, which can be used to prevent psychomotor seizures.
  • The compound is the grade of analytical standard.
  • It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-003-43206