Learn More
Cayman Chemical PhenformInd5hydrochLRIde 10mg
SDP
Supplier:  Cayman Chemical 3634110 MG
Encompass_Preferred
An internal standard for the quantification of phenformin by GC- or LC-MS
Catalog No. 50-291-2229