Learn More
Medchemexpress LLC Phenibut hydrochloride | 3060-41-1 | MFCD00075860 | >98.0% | 215.68 g/mol | C10H14ClNO2 | 100 G

Supplier: Medchemexpress LLC HYW041333100G
Phenibut hydrochloride is a GABAB receptor agonist supplied as a white to off-white powder for research use. It is used in pharmacology and neuroscience studies investigating GABAergic signaling, anxiolytic effects, and cognition-related research.
- Purity ≥98.0%.
- CAS number 3060-41-1.
- Molecular formula C10H14ClNO2.
- Molecular weight 215.68 g/mol.
- Physical form: white to off-white powder.
- Available in laboratory package sizes up to 100 g.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.