Learn More
Medchemexpress LLC Phenol, 2,4,6-triiodo- | 609-23-4 | MFCD00002179 | 98.3% | 471.80 g/mol | C6H3I3O | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW073687
Encompass_Preferred
2,4,6-Triiodophenol is a small-molecule research compound described as an orally active leukotriene B4 (LTB4) synthesis inhibitor and reported to induce apoptosis in mouse blastocysts. It is supplied for laboratory research in solid and solution forms, including a 10 mM solution in DMSO.
  • orally active leukotriene B4 (LTB4) synthesis inhibitor.
  • reported to induce apoptosis in mouse blastocysts.
  • available as a solid and as a 10 mM solution in DMSO.
  • high purity suitable for research applications.
  • chemical formula C6H3I3O and molecular weight 471.80 g/mol.
Catalog No. 50-002-09082