Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenol, 2-bromo-4-methyl- | 6627-55-0 | MFCD00002151 | 99.3% | 187.03 g/mol | C7H7BrO | 500 G
SDP

Supplier:  Medchemexpress LLC HYW016153500G

Encompass_Preferred

2-Bromo-4-methylphenol (CAS 6627-55-0) is a brominated phenol used as a biochemical reagent and a building block in organic synthesis and life-science research. It is supplied as a colorless to light-yellow liquid with high purity, and includes solubility and storage data for both in vitro and in vivo applications.

  • High purity suitable for research and synthesis.
  • Molecular formula C7H7BrO; molecular weight 187.03 g/mol.
  • Soluble in DMSO (100 mg/mL) and formulatable for in vivo dosing.
  • Stable with defined storage conditions and shelf life.
  • Available in laboratory pack sizes up to 500 g for scale-up.

Catalog No. 50-003-02276


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.