Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenol, 3,4,6-trichloro-2-[(2,3,5-trichloro-6-hydroxyphenyl)sulfonyl]- | 1638-41-1 | MFCD00397032 | 99.0% | 456.94 g/mol | C12H4Cl6O4S | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1124015MG

Encompass_Preferred

p38 MAP Kinase Inhibitor IV is an ATP-competitive inhibitor selective for p38α and p38β MAPKs, used as a biochemical tool to probe p38 signaling. It has IC50 values of 0.13 μM (p38α) and 0.55 μM (p38β), and is supplied as a high-purity solid for research applications.

  • ATP-competitive inhibitor with defined IC50 values for mechanistic studies.
  • High purity (99.0%) for reliable biochemical and cellular assays.
  • Solid powder form for convenient storage and handling.
  • Recommended storage conditions provided to preserve stability.
  • Molecular weight and formula supplied for analytical and formulation work.

Catalog No. 50-002-64143


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.