Learn More
Medchemexpress LLC Phenol, 3,5-bis(1,1-dimethylethyl)- | 1138-52-9 | MFCD00008829 | 100.0% | 206.33 g/mol | C14H22O | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW041080
Encompass_Preferred
3,5-Di-tert-butylphenol is a substituted phenolic research reagent (CAS 1138-52-9) used in antifungal and anti-biofilm studies and as a chemical building block for analytical and synthetic applications. It is supplied as a high-purity solid and as ready-to-use DMSO solutions, and is provided with supporting documentation for research use.
  • High purity, 99.98% assay reported on the COA.
  • Available as solid and as 10 mM solutions in DMSO.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
  • Offered in multiple package sizes, including 25 g and larger bulk quantities.
  • Molecular formula C14H22O; molecular weight 206.33 g/mol.
Catalog No. 50-002-09669