Learn More
Medchemexpress LLC Phenol, 4-(1H-imidazol-1-yl)- | 10041-02-8 | 160.18 | 10 G
Supplier: Medchemexpress LLC HYW00035710G
4-(Imidazol-1-yl)phenol is a highly potent signal enhancer used in horseradish peroxidase (HRP)-luminol chemiluminescence (CL) immunoassays. It is intended for research use only and not sold to patients.
- Highly potent signal enhancer
- Used in HRP-luminol chemiluminescence immunoassays
- For research use only