Learn More
Medchemexpress LLC Phenol, 4-(2-aminoethyl)-2-methoxy | 554-52-9 | MFCD00060621 | ≥97.0% | 167.21 | C9H13NO2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY103638A500MG
Encompass_Preferred
3-Methoxytyramine (3-O-methyl dopamine) is an extracellular metabolite of dopamine used in biochemical and neuromodulator research. It is supplied as a solid (yellow to brown) and as a 10 mM solution in DMSO, with supporting analytical and safety documentation for research use.
  • Research-grade dopamine metabolite for biochemical studies.
  • Available as solid and as a 10 mM solution in DMSO.
  • Purity ≥97.0% with certificate of analysis available.
  • Pack sizes include 100 mg, 250 mg, 500 mg, 1 g, and 5 g.
  • Safety data sheet and datasheet PDFs provided for handling and analysis.
Catalog No. 50-002-46721