Learn More
Medchemexpress LLC Phenol, 4,4′-(2-pyridinylmethylene)bis- | 603-41-8 | MFCD00023496 | 99.0% | 277.32 g·mol⁻¹ | C18H15NO2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1316375MG
Encompass_Preferred
Desacetyl bisacodyl is the active metabolite of the laxative bisacodyl, supplied as a solid research reagent for pharmacological and metabolic studies. It induces epithelial chloride secretion in rodent colon and rectum and is intended for use as a reference standard or analytical standard in laboratory experiments.
  • High purity (99.0%) suitable for analytical and pharmacological assays.
  • Defined molecular weight and formula for precise calculations.
  • Available in small pack sizes for trace-level experiments.
  • Supplied as a solid for stable storage and handling.
  • Used as an active metabolite reference in metabolic and physiological studies.
Catalog No. 50-002-62176