Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenol, 4-(6-methyl-1,2,4,5-tetrazin-3-yl)- | 58884-35-8 | 97.53% | C9H8N4O | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHY141274

Encompass_Preferred

4-(6-Methyl-1,2,4,5-tetrazin-3-yl)phenol is an alkyl chain-based PROTAC linker designed for the synthesis of PROTACs. These Proteolysis Targeting Chimeras consist of two different ligands connected by a linker, targeting an E3 ubiquitin ligase and a specific target protein. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.

  • Appearance: Pink to red solid
  • Solubility in DMSO: 100 mg/mL
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month
  • Related classification: Click Chemistry (Tetrazine)
  • Relevant disease areas: Cancer, Cancer Targeted Therapy

Catalog No. 50-003-52751


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.