Learn More
Medchemexpress LLC Phenol, 4-amino-3-methyl- | 2835-99-6 | ≥98.0% | 123.16 | 50 G
Supplier: Medchemexpress LLC HYW01282450G
4-Amino-m-cresol is recognized for its role as a soil urease inhibitor and is categorized as a biochemical assay reagent. This compound has been documented to actively suppress the activity of urease in soil environments.
- Inhibits soil urease activity.
- Intended for research applications.
- For research use only.