Learn More
Medchemexpress LLC Phenol, 4-ethoxy- | 622-62-8 | MFCD00002334 | 99.9% | 138.16 g/mol | C8H10O2 | 10 G

Supplier: Medchemexpress LLC HYW01578610G
4-Ethoxyphenol is a phenolic aromatic ether used in chemical research and assay development. Provided as a solid with high purity, it functions as a reagent or analytical standard in enzymatic and microbiological studies. Handle and store according to recommended conditions to preserve stability and purity.
- High purity (99.86%) suitable for analytical and research use.
- Solid form, off-white to light brown, easy to weigh and dissolve.
- Molecular weight 138.16 g/mol; molecular formula C8H10O2.
- Available in laboratory pack sizes for small-scale studies.
- Recommended storage: powder at -20°C for long-term, 4°C for short-term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.