Learn More
Medchemexpress LLC Phenol, 4-(phenylmethoxy)- | 103-16-2 | 99.9% | 200.24 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY30272
Encompass_Preferred
Monobenzone is a potent skin depigmenting agent that induces depigmentation and shows potential for vitiligo research. It is also a potent inhibitor of RNR (Ribonucleotide reductase) enzyme activity by targeting RRM2 protein, demonstrating significant anti-leukemia efficacy in vitro and in vivo. It inhibits acute myeloid leukemia (AML) cell proliferation and DNA synthesis, and induces cell cycle arrest and apoptosis.
  • Potent skin depigmenting agent.
  • Induces depigmentation and shows potential for vitiligo research.
  • Potent inhibitor of RNR enzyme activity.
  • Exhibits significant anti-leukemia efficacy in vitro and in vivo.
  • Inhibits AML cell proliferation and DNA synthesis.
  • Induces cell cycle arrest and apoptosis.
Catalog No. 50-004-03301