Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenol-amido-C1-PEG3-N3 | 1096439-18-7 | 99.9% | 324.33 g/mol | C14H20N4O5 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1288355MG

Encompass_Preferred

Phenol-amido-C1-PEG3-N3 is an azido-functionalized PEG-based PROTAC linker used as a building block in targeted protein degradation research and medicinal chemistry. Provided as a liquid with a light brown appearance, it offers high purity and defined molecular properties suitable for synthesis workflows and click-chemistry conjugation.

  • High purity approx. 99.9%.
  • Molecular formula C14H20N4O5, molecular weight 324.33 g/mol.
  • Azido functional group enables CuAAC and SPAAC click reactions.
  • Supplied as a liquid, color light brown to brown.
  • Storage: pure form at -20°C; in solvent at -80°C for extended stability.
  • Available in small research pack sizes for medicinal chemistry workflows.

Catalog No. 50-002-62180


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.