Learn More
Medchemexpress LLC Phenol-amido-C1-PEG3-N3 (PROTAC Linker 21) | 1096439-18-7 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12883550MG
Encompass_Preferred
Phenol-amido-C1-PEG3-N3 is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Alkyne groups. It can also engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent with an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reactions
  • Undergoes strain-promoted alkyne-azide cycloaddition reactions
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-38772