Learn More
Sigma Aldrich Fine Chemicals Biosciences Phenol BioUltra for molecu1L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 776071L
Encompass
saturated with 10 mM TRIS buffer pH 8.0 and 1 mM EDTA
Catalog No. 50-290-7569