Learn More
Hach Company Phenol Reagent Powder Pillows (for nonextraction method), pk/100
Supplier:  Hach Company 2481569
Encompass
For determination of phenols by the 4-Aminoantipyrine Method
Catalog No. 50-199-6053