Learn More
Selleck Chemical LLC Phenol Red E7437-1G
SDP
Supplier:  Selleck Chemical LLC E74371G
Encompass_Preferred
Phenol red is a pH indicator dye commonly used in cell culture media where it changes color from yellow to red based on pH levels It also participates in redox reactions particularly in the presence of enzymes like myeloperoxidase influencing cellular oxidative states without directly causing significant toxicity
Catalog No. 50-000-41064