Learn More
Sigma Aldrich Fine Chemicals Biosciences Phenol solution BioReagent, Saturated with 0.1 M citrate buffer, pH 4.3 +/- 0.2, for molecular biology | 108-95-2 | MFCD00002143 | 100ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P4682100ML
Encompass
Phenol solution BioReagent, Saturated with 0.1 M citrate buffer, pH 4.3 +/- 0.2, for molecular biology | Mol Wt: 94.11 | 108-95-2 | MFCD00002143 | 100ML
Catalog No. 50-185-7380