Learn More
Hach Company Phenolphthalein Indicator Solution, 5 g/L, 1 L (APHA)
Supplier:  Hach Company 16253
Encompass
pH indicator for use in titrations. Color change: colorless to pink at ~pH 8.3. 1 liter bottle.
Catalog No. 50-199-4186