Learn More
Aqua Solutions Phenol Red 0.04% w/v Solution (pH 6.8 yellow - 8.2 red) (500ML)
SDP
Supplier:  Aqua Solutions 6700500ML
Encompass_Preferred
Phenol Red 0.04% w/v Solution (pH 6.8 yellow - 8.2 red) (500ML)
Catalog No. NC9077589
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov