Learn More
Phenomenex Inc Phenomenex SEC Column, 1.8 μm, SEC-2, 150 x 4.6 mm
Supplier: Phenomenex Inc PART: 00F4769E0
Size-exclusion liquid chromatography column built with 1.8 μm thermally modified, fully porous particles and 150 Å pores for high-efficiency separation and reliable analysis of low molecular weight biomolecules (approx. 1 k-450 kDa). The column is rated for UHPLC pressures and offers broad pH stability for robust method work.
- High-efficiency 1.8 μm fully porous particles for improved resolution and peak shape.
- Optimized 150 Å pore size for proteins, peptides, and low-MW analytes.
- Suitable for LMW separations in the 1 k-450 kDa range.
- Titanium hardware reduces secondary interactions and minimizes carryover.
- Wide pH stability (2.5-7.5) for flexible mobile phase selection.
- High-pressure rating (up to 7,000 psi) compatible with UHPLC systems.
- Thermally modified particle surface for enhanced stability and reproducibility.
Description
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.