Learn More
Phenomenex Inc Phenomenex SEC Column, SEC-3, 1.8 μm, 50 x 2.1 mm
Supplier:  Phenomenex Inc PART 00B4772AN
Encompass
High-efficiency 1.8 μm size-exclusion LC column optimized for separation and characterization of proteins and other macromolecules. The SEC-3 chemistry and 300 Å pore structure provide improved resolution for low- to mid-molecular-weight analytes while supporting UHPLC-compatible pressures and temperature-controlled analyses.
  • 1.8 μm particle size for enhanced chromatographic efficiency.
  • SEC-3 stationary phase optimized for size exclusion separations.
  • 50 x 2.1 mm column dimensions for fast, high-resolution analyses.
  • 300 Å pore size suited to protein and macromolecule separations.
  • Up to 600 bar maximum operating pressure for UHPLC compatibility.
  • pH stability range 2-8 for broad mobile phase options.
  • Maximum operating temperature 80 °C to support temperature-controlled runs.
  • Designed for protein characterization, aggregate and purity assessment.
Catalog No. NC3842364