Learn More
Medchemexpress LLC HY-13721 5mg Medchemexpress, Phenoxodiol CAS:81267-65-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY137215MG
Encompass_Preferred
Medchemexpress, HY-13721 5mg Phenoxodiol CAS:81267-65-4 Phenoxodiol, a synthetic analog of Genestein, activates the mitochondrial caspase system, inhibits XIAP (an apoptosis inhibitor), and sensitizes the cancer cells to Fas-mediated apoptosis. This agent also inhibits DNA topoisomerase II by stabilizing the cleavable complex. Phenoxodiol induces cell cycle arrest in the G1/S phase of the cell cycle and upregulates p21WAF1 via a p53 independent manner. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9886