Learn More
Medchemexpress LLC Phenoxymethylpenicillin | 87-08-1 | 98.9% | 100 MG

Supplier: Medchemexpress LLC HYB0975A100MG
Penicillin V (Phenoxymethylpenicillin) is a potent and orally active antibiotic. It exhibits antibacterial activity against Streptococci, Clostridium difficile, and Staphylococcus aureus. This compound is being researched for its potential use in otitis, sinusitis, pharyngitis, and tonsillitis.
- Potent and orally active antibiotic
- Exhibits antibacterial activity against Streptococci
- Exhibits antibacterial activity against Clostridium difficile
- Exhibits antibacterial activity against Staphylococcus aureus
- Researched for potential use in otitis, sinusitis, pharyngitis, and tonsillitis
- Solid, white to off-white appearance
- Molecular weight of 350.39
- Chemical formula: C16H18N2O5S
- Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
- Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.