Learn More
Medchemexpress LLC HY-16009 5mg Medchemexpress, (+)-Phenserine CAS:116839-68-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY160095MG
Encompass_Preferred
Medchemexpress, HY-16009 5mg (+)-Phenserine CAS:116839-68-0 (+)-Phenserine is a novel selective cholinesterase noncompetitive inhibitor with an IC50 of 45.3 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1860