Learn More
Cayman Chemical PhentolamIn-d4hydrochlorId 1mg
SDP
Supplier:  Cayman Chemical 294171 MG
Encompass_Preferred
An internal standard for the quantification of phentolamine by GC- or LC-MS
Catalog No. 50-298-9116