Learn More
Selleck Chemical LLC Phentolamine Mesylate-50mg
SDP
Supplier:  Selleck Chemical LLC S203850MG
Encompass_Preferred
Phentolamine Mesylate(Phentolamine methanesulfonate) is a reversible and nonselective alpha-adrenergic receptor antagonist, used for the prevention or control of hypertensive episodes.
Catalog No. 50-238-9734