Learn More
Cayman Chemical PhentolamInmesylate 500mg
SDP
Supplier:  Cayman Chemical 16135500 MG
Encompass_Preferred
A reversible antagonist of α-adrenergic receptors, non-specifically binding all α1- and α2-adrenoceptors with nanomolar affinities; rapidly and transiently increases local blood flow
Catalog No. 50-296-8703