Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc PHENYL 20UM 120A 500X20MM
SDP

Supplier:  YMC America, Inc PH12S215020WT

Encompass_Preferred

Phenyl 20Um; 120A; 500 X 20mm

Catalog No. 50-900-05930


May include imposed supplier surcharges.
Only null left
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.