Learn More
Selleck Chemical LLC Phenyl-ac-Gly-OH
SDP
Supplier:  Selleck Chemical LLC S605125MG
Encompass_Preferred
Phenyl-ac-Gly-OH (N-(2-Phenylacetyl)glycine 2-Phenylacetyl glycine Phenaceturic acid Phenacetylglycine) is one of acyl glycines that are normally minor metabolites of fatty acids
Catalog No. 50-000-03410