Learn More
Medchemexpress LLC Phenyl benzoate | 93-99-2 | 99.9% | 198.22 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00982310G
Encompass_Preferred
Phenyl benzoate is a benzoate ester obtained by the formal condensation of phenol with benzoic acid. It acts as a chloride transport blocker, inhibiting Cl--dependent Glu accumulation into vesicles, and can also be used as a preservative in cosmetic products.
  • Benzoate ester derived from phenol and benzoic acid
  • Acts as a chloride transport blocker
  • Inhibits Cl--dependent Glu accumulation
  • Can be used as a preservative in cosmetic products
Catalog No. 50-004-52524