Learn More
Medchemexpress LLC Phenyl-boronic acid-d5 | 215527-70-1 | MFCD02093711 | 126.96 | C6H2D5BO2 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0676155G
Encompass_Preferred
Phenyl-boronic acid-d5 is the deuterium-labeled analogue of phenylboronic acid used as an isotope reagent in analytical and synthetic chemistry. It is typically used as a mass spectrometry standard, tracer, or building block in cross-coupling reactions.
  • Deuterium-labeled reagent for analytical applications.
  • Used as an internal standard for mass spectrometry.
  • Suitable for isotopic tracing and synthetic chemistry.
  • Solid, stable under normal laboratory storage conditions.
Catalog No. 50-002-10558