Learn More
Sigma Aldrich Fine Chemicals Biosciences Phenyl isothiocyanate for
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 78780100ML
Encompass
Phenyl isothiocyanate is an aromatic isothiocyanate. It participates in dehydration reactions of alcohols. It is widely used for synthesis of various biologically important heterocyclic compounds.
Catalog No. 50-289-8284