Learn More
Sigma Aldrich Fine Chemicals Biosciences Phenyl isothiocyanate Sigm
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P103410ML
Encompass
Phenyl isothiocyanate belongs to the class of aromatic compounds with isothiocyanate groups. It is typically used as a precolumn derivatization agent for the formation of phenylthiocarbamyl amino acids for their subsequent determination using chromatography techniques.
Catalog No. 50-289-1487